Random mutagenesis kits geneMorph II
Fournisseur: AGILENT
200552 200550 200552
STRA200552EA 0 EUR InStock
STRA200552 AGLS200550 AGLS200552
Random mutagenesis kits geneMorph II
Réactifs pour acides nucléiques Cloning Kits and Ligation
The GeneMorph II random mutagenesis kits have been formulated to produce a more uniform mutational spectrum when performing error prone PCR to achieve random mutagenesis.
  • Superior yields over a wide target range - 0,1 to 6 kb
  • Easily change mutation frequency by altering concentration of input template
  • Efficient mutagenesis rates of 1 to 16 bases per kb
Other methods or commercially available kits that rely on Taq DNA polymerase for random mutagenesis have shown biases towards mutating A’s and T’s more frequently than G’s and C’s, which undoubtedly skews representation of random mutant libraries.
GeneMorph II kits utilise Mutazyme II DNA polymerase, which is a novel error prone PCR enzyme blend with equivalent mutation rates at A’s and T’s vs. G’s and C’s. Additionally, to address the need for efficient and flexible cloning methods while delivering this less biased mutational spectrum. The GeneMorph II random mutagenesis kit takes error-prone PCR to the next level by combining mutational spectrums of both our Mutazyme DNA polymerase and Taq DNA polymerase.
GeneMorph II EZClone Domain Mutagenesis Kit offers an easy and fast cloning method to perform targeted random mutagenesis on protein domains and promoter elements and eliminates the need to have specific restriction sites.
Attention: For research use only. Not for use in diagnostic procedures.
Collapse All
Désignation No. of reactions Cdt Numéro de catalogue VWR Quantité
GeneMorph II EZClone domain mutagenesis kit 10 1 kt AGLS200552
GeneMorph II random mutagenesis kit 30 1 kt AGLS200550
claquement
Veuillez sélectionner l'article
Entrer les détails
SPÉCIFICATIONS
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité