InterPlay adenoviral TAP system
Fournisseur: AGILENT
240101
AGLS240101EA 783 EUR InStock
AGLS240101
InterPlay adenoviral TAP system
Vecteurs, plasmides et librairies
The InterPlay Mammalian TAP system allows you to recover interacting proteins from mammalian cells.
  • Protease digestion unnecessary to recover intact interacting protein partners since SBP and CBP elute easily from their affinity resins
  • Exceptionally pure protein for downstream analysis
This method is based on expression of a protein of interest fused to two affinity tags: a streptavidin binding peptide (SBP) and a calmodulin binding peptide (CBP). Tandem affinity purification yields your tagged protein and interacting proteins using gentle washing and small molecule elution conditions.
Attention: For research use only. Not for use in diagnostic procedures.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité