Proteinase K
Fournisseur: AGILENT
300140
AGLS300140EA 212 EUR InStock
AGLS300140
Proteinase K
Enzymes
Proteinase K is a broad spectrum protease from Tritirachium album that is ideal for preparing chromosomal DNA.
  • For general digestion of protein in biological samples
  • Lyophilised powder with specific activity >30 units/mg at 37 °C
  • Size: 100 mg
It cleaves peptide bonds in which carbonyl groups are surrounded by aromatic, hydrophobic, or aliphatic amino acids.
Used for the following applications: pulsed field electrophoresis, chromosomal DNA for protein fingerprinting, and nuclease removal from DNA and RNA preparations.
Attention: For research use only. Not for use in diagnostic procedures.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité