Multiple affinity removal column HSA/IgG
Fournisseur: AGILENT
5188-8826
AGLS5188-8826EA 1420 EUR InStock
AGLS5188-8826
Multiple affinity removal column HSA/IgG
Colonnes pour chromatographie
The multiple affinity removal LC column – human albumin/IgG is designed to chromatographically remove the two most interfering high-abundant proteins (albumin and IgG) from human samples (such as plasma, serum, urine or cerebrospinal fluid).
  • Serum injection volume is 100 µl per run for the 4,6 mmid column
  • Optimise sample recovery by utilising rapid, single injection protocols and optimised buffers
  • Column is compatible with most FPLC or HPLC pumps or instruments
Removal of albumin and IgG improves the subsequent LC/MS and electrophoretic analysis of the sample by effectively expanding the dynamic range of the analysis.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité