Taq extender PCR additive
Fournisseur: AGILENT
600148
AGLS600148EA 291 EUR InStock
AGLS600148
Taq extender PCR additive
Réactifs pour acides nucléiques qPCR/RT-qPCR Enzymes and Kits
Improves reliability and yield of conventional Taq-based PCR amplifications.
  • Improves length, yield, and reliability of PCR
  • Enhances efficiency of template extension by Taq DNA polymerase
  • Improves amplification of difficult targets
Taq Extender PCR additive improves the reliability and yield of conventional Taq-based PCR amplifications. Taq Extender additive increases the efficiency of Taq DNA polymerase extension reactions during each cycle of PCR by reducing mismatch pausing, resulting in a greater percentage of completed extension reactions.
Attention: For research use only. Not for use in diagnostic procedures.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité