miRNA reference kit
Fournisseur: AGILENT
750700
AGLS750700EA 654 EUR InStock
AGLS750700
miRNA reference kit
Réactifs pour acides nucléiques RNAi
The Universal Human miRNA Reference RNA is an ideal reference control for miRNA microarray or miRNA-targeted QRTPCR experiments.
  • Contains extensive range of miRNAs for genome-wide detection
  • Includes mRNA, large intergenic non-coding RNA, and piwi-interacting RNA in addition to miRNA
  • Non-synthetic, not limited to previously characterized miRNA species
  • Carefully screened for contaminating genomic DNA
  • Created in extremely large lots with stringent QC procedures, ensuring long-term availability of consistent reference RNA material
The Universal Human miRNA Reference RNA may also be used as an optimization or standardization reagent for these or other applications aimed at human miRNA analysis.
It is composed of total RNA from nine human tissues or cell lines specifically selected and formulated for optimal miRNA representation.
Attention: For research use only. Not for use in diagnostic procedures.
Collapse All
Désignation Cdt Numéro de catalogue VWR Quantité
Universal miRNA reference kit, 10 ug 1 KIT AGLS750700
claquement
Veuillez sélectionner l'article
Entrer les détails
SPÉCIFICATIONS
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité