Plasmid DNA Kit
Fournisseur: AGILENT
DNF-940-K0500
AGLSDNF-940-K0500EA 411 EUR InStock
AGLSDNF-940-K0500
Plasmid DNA Kit
Systèmes d'électrophorèse Capillaires pour électrophorèse
Quality control analysis of supercoiled and linear plasmid DNA for downstream applications.
  • Reliable sizing- optimised ladder for the accurate sizing of supercoiled plasmids
  • Concentration range - 0.1 to 1 ng/ul
  • Fast analysis - QC results in only 30 minutes
  • Accurate sizing - sizing range from 2,000 to 10,000 bp for supercoiled plasmids
The plasmid DNA kit is used in the analysis of supercoiled and linear plasmid DNA. The ladder provided with this kit is optimised for the accurate sizing and relative quantification of supercoiled plasmids between 2,000 and 10,000 bp. Linearised plasmids can be evaluated for quality and relative quantification, however only comparative sizing is possible.
Attention: For research use only. Not for use in diagnostic procedures.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité