Ultra Sensitivity RNA Kit
Fournisseur: AGILENT
FP-1201-0275
AGLSFP-1201-0275EA 1620 EUR InStock
AGLSFP-1201-0275
Ultra Sensitivity RNA Kit
Systèmes d'électrophorèse Capillaires pour électrophorèse
Quality control analysis of mRNA and total RNA.
  • Ultra-sensitive detection - The ultra sensitivity RNA kit provides detection of RNA at concentrations as low as 2,5 pg/µl for total RNA and 15 pg/µl for mRNA
  • Quality metrics - The RNA Quality Score (RQN) provides objective analysis of total RNA samples and percent ribosomal contamination for mRNA samples
  • Reliable sizing - The ultra sensitivity RNA kit provides accurate sizing through 6000 nt
  • Low sample volume - Only 2 µl of sample is required, minimising sample loss to RNA QC steps
  • Two separation methods - Optimised separation methods for total RNA (US Total RNA method) and mRNA (US mRNA method)
The ultra sensitivity RNA kit is used for the assessment of low concentration RNA with as low as 2,5 pg/µl. Two separation methods are optimised for the analysis of total RNA and mRNA QC. Both sample types require quality control steps in NGS library preparation and gene expression analysis workflow.
ProSize data analysis software simplifies the RNA QC process by providing an RNA Quality Score (RQN) for total RNA and a percent ribosomal RNA contamination for mRNA samples. These quality metrics are automatically calculated by ProSize software allowing for objective analysis of samples during RNA QC.
Attention: For research use only. Not for use in diagnostic procedures.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité