Bravo NGS
Fournisseur: AGILENT
G5573AA
AGLSG5573AAEA 0 EUR InStock
AGLSG5573AA
Bravo NGS
Instruments de préparation d'échantillons
Automate genomics workflows for NGS without sacrificing quality, in a small footprint.
  • Quickly automate with ready-to-use protocols and pre-developed methods for Agilent SureSelect, Agilent HaloPlex, Illumina TruSeq, NuGEN Ovation, KAPA HTP, PacBio SMRTbell, NEBNext, and more
  • Easy to use interface requires no programming expertise to run your automated methods
  • Consistent data quality allows you to increase your throughput, with consistent results every time
  • Prepare up to ten times more samples in the same time
  • Open platform allows you to automate more NGS or genomics methods with confidence
  • The optional on-deck thermal cycler (ODTC) is a compact and configurable accessory on the Bravo NGS system which increases walk-away time and simplifies workflows
The Agilent Bravo NGS is a flexible and compact automated liquid handling platform that streamlines routine genomic workflows, including library preparation and target enrichment for next generation sequencing (NGS). Agilent VWorks automation control software allows users to quickly start the system and run preprogrammed protocols with confidence, while the open platform provides the flexibility to serve a wide range of applications.
The Bravo NGS offers the advantage of increased throughput without affecting data quality, and frees researchers from performing time-consuming and repetitive tasks. The integrated on-deck thermal cycler (ODTC) further increases walkaway time and greatly simplifies the NGS sample preparation workflow. The compact design of the Agilent Bravo automated liquid handing platform makes it an excellent choice for laboratories with space constraints.
Attention: For Research Use Only. Not for use in diagnostic procedures.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité