ZAG DNA Analyser System
Fournisseur: AGILENT
M5320AA
AGLSM5320AAEA 0 EUR InStock
AGLSM5320AA
ZAG DNA Analyser System
Systèmes d'électrophorèse Capillaires pour électrophorèse
Parallel capillary electrophoresis system for automated high-throughput DNA fragment analysis.
  • High separation resolution - reliably distinguish and size DNA fragments within 3 bp of each other
  • High throughput - process 48, 96-well sample trays in 24 hours. Broad sizing range - size fragments between 35 bp and 20,000 bp through a variety of available DNA analysis kits
  • Fast separation times - samples can be separated in as little as 20 minutes
  • Intuitive data analysis - analytical software can process separation data automatically after the run and generate a report in multiple formats
  • desktop PC supplied with a comprehensive 3-year warranty through HP
The Agilent ZAG DNA analyser system relieves bottlenecks in automated high-throughput DNA fragment analysis workflows. Using parallel capillary electrophoresis, the ZAG DNA Analyser system can process thousands of DNA fragment samples a day, eliminating bottlenecks associated with agarose gels.
This system is ideal for a range of DNA fragment applications, from basic PCR fragment analysis to microsatellite detection. Minimal sample preparation, automated separation of samples, and the option for continuous operation help keep your workflow moving.
Attention: For research use only. Not for use in diagnostic procedures.
Commander maintenant
COMMANDE
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité