Oligo Pro II Parts and Accessories
Fournisseur: AGILENT
P60-22NM
AGLSP60-22NMEA 21.4 EUR InStock
AGLSP60-22NM
Oligo Pro II Parts and Accessories
Systèmes d'électrophorèse Capillaires pour électrophorèse
If you need spare parts and accessories for the Oligo Pro II system search our spare parts and accessory portfolio page. Find out more here.
  • Gel bottle cap and waste bottle cap replacement - modified for liquid tubing to be used with the Oligo Pro II system
  • Tray carrier adaptor - molded plate adaptor designed to hold semi-skirted PCR plate in the drawer location
  • High-quality products for low instrument downtime and best total cost of ownership
Obtain spare parts and accessories specifically designed for the Oligo Pro II system to ensure continued high level of system operation for the quality control of oligonucleotides.
The selection of spare parts includes gel bottle cap replacement, waste bottle replacement cap, carrier for the buffer and waste trays as well the open waste tray specific designed for the Oligo Pro II system.
Attention: For research use only. Not for use in diagnostic procedures.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité