Thermometer similar to ASTM 1C, solid stem, -20+150:1C, white backed, red special liquid, 317-327x6-7mm, immersion 76mm, top finish round, durable pigment, suitable for government verification coated with PTFE 1 * 1 Pce
Fournisseur: AMARELL, ARNO
Thermometer similar to ASTM 1C, solid stem, -20+150:1C, white backed, red special liquid, 317-327x6-7mm, immersion 76mm, top finish round, durable pigment, suitable for government verification coated with PTFE 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Thermometer similar to ASTM 1C, solid stem, -20+150:1°C, white backed, red special liquid, 317-327x6-7mm, immersion 76mm, top finish round, durable pigment, suitable for government verification coated with PTFE 1 * 1 Pce AMARA300010-FLPTFE 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou