Apron, sleeveless, AlphaTec® 4000, Model 212
Fournisseur: ANSELL HEALTH CARE
GR40W-00212-00
ANSEGR40W-00212-00EA 1320 EUR InStock
ANSEGR40W-00212-00
Apron, sleeveless, AlphaTec® 4000, Model 212
Vêtements à usage unique
Innovative multi-layer chemical barrier technology.
  • Exceptional protection, over 200 chemicals permeation tested, including chemical warfare agents
  • Silicone-free
Aprons feature tie fastening to the waist with a 1000 mm long tie fastening.
Recommended for contaminated environments, chemical handling, filling/blending/charging raw materials, transferring liquids and solids, opening and draining pumps valves or lines, loading and unloading of proces equipment, raw material sample collection, concentration mixing and palletisation, Hazmat response, and large pipe assembly/disassembly.
Certifications: Type PB [3]-B; Type PB [4]-B; EN 14126; Category III
Conditionnement: Packaged 100 pieces/carton.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité