Chemical resistant sleeve protectors, AlphaTec® 4000, Model 600
Fournisseur: ANSELL HEALTH CARE
GR40W-00600-00
ANSEGR40W-00600-00EA 1010 EUR InStock
ANSEGR40W-00600-00
Chemical resistant sleeve protectors, AlphaTec® 4000, Model 600
Vêtements à usage unique
These sleeve protectors are made from a multi-layer chemical barrier technology that provides an exceptional barrier against a wide range of organic and inorganic chemicals and biological agents.
  • Elasticated at both ends and welded seams
  • Designed to protect
  • Silicone free
Typical applications include: Viral contaminated areas (including Avian Influenza), biological protection, mining, fire and rescue, sewage purification installations.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité