Chemical resistant boot covers, AlphaTec® 2000, Model 406
Fournisseur: ANSELL HEALTH CARE
WH20B-00406-00
ANSEWH20B-00406-00EA 378 EUR InStock
ANSEWH20B-00406-00
Chemical resistant boot covers, AlphaTec® 2000, Model 406
Vêtements à usage unique
Superior breathable microporous laminate technology.
  • Proven protective barrier to low concentration liquid chemicals, liquid, and particulate biological hazards
  • Comfortable moisture vapor permeable fabric helps reduce the risk of heat stress
  • Optimised fit, improving wearer comfort and safety
  • Low Linting
Boot covers feature tie fastening, elastic to top of boot, and bound seams. Covers are manufactured from non-woven microporous polyethylene laminate material.
Recommended for cleanroom preparation and cleaning, production line support and maintenance, plant and machine maintenance, and veterinary services.
Certifications: Category I
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité