Sleeve protectors, AlphaTec® 2000, model 600
Fournisseur: ANSELL HEALTH CARE
WH20B-00600-00
ANSEWH20B-00600-00EA 623 EUR InStock
ANSEWH20B-00600-00
Sleeve protectors, AlphaTec® 2000, model 600
Vêtements à usage unique
These sleeve protectors are made from a non woven fabric with microporous PE film. The fabric provides an effective liquid and particle barrier combined with a high water vapour transmission rate from inside to outside. Ideal for a wide range of industrial applications.
  • Elasticated at both ends
  • Bound seams
  • Lightweight and comfortable
  • Excellent liquid penetration resistance and barrier to fine particulates
  • Silicone free: Critical in paint spraying applications
  • Anti-static: Tested according to EN 1149-5
Typical applications include: Pharmaceutical industries, agriculture, cleanrooms, paint spraying, crime scene investigation, veterinary services.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité