Chemical Resistant Cape Hood, AlphaTec® 2500, model 507
Fournisseur: ANSELL HEALTH CARE
WH25B-00507-00
ANSEWH25B-00507-00EA 1050 EUR InStock
ANSEWH25B-00507-00
Chemical Resistant Cape Hood, AlphaTec® 2500, model 507
Vêtements à usage unique
These hoods are made from a durable microporous PP laminate with a non woven inner layer. The fabric provides an excellent barrier to chemical spray and infective agents.
  • Balaclava style: covers part of shoulders
  • Velcro fastening to front
  • Bound seams
  • Low linting: reduced risk of contamination in critical areas
Typical applications include: Viral contaminated areas (including Avian Influenza), biological protection, emergency medical response, medical research, chemical and pharmaceutical industries, low pressure industrial, cleaning, industrial paint spraying, nuclear industry.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité