A247118-96
ANTIA247118-96EA
635
EUR
InStock
ANTIA247118-96
Human Humanin/MT-RNR2 ELISA Kit
Tests
Tests ELISA
Human Humanin/MT-RNR2 ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Humanin/MT-RNR2 in serum, plasma, tissue homogenates, and other biological fluids.
- Ready-to-use ELISA kit
- Detection range: 0,146 to 600 pg/ml
- Sensitivity: 18,75 pg/ml
- Sample type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%
Human Humanin/MT-RNR2 ELISA kit employs the competitive enzyme immunoassay technique for the quantitative measurement of human Humanin/MT-RNR2 in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with Humanin/MT-RNR2 antigen. During the incubation, Humanin/MT-RNR2 present in the samples or standards competes with the fixed amount of immobilised Humanin/MT-RNR2 for binding sites on the Biotinylated Anti-Humanin/MT-RNR2 antibody. The more Humanin/MT-RNR2 present in a sample or standard, the less Biotinylated Anti-Humanin/MT-RNR2 antibody that binds to the plate. Following incubation, unbound Biotinylated Anti-Humanin/MT-RNR2 antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of Humanin/MT-RNR2 present in each sample or standard. The concentration of Humanin/MT-RNR2 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.
Informations de commande: This is a ready-to-use ELISA kit that contains: pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.