Canine Triiodothyronine/T3 ELISA Kit
Fournisseur: ANTIBODIES.COM
A302563-96
ANTIA302563-96EA 710 EUR InStock
ANTIA302563-96
Canine Triiodothyronine/T3 ELISA Kit
Tests Tests ELISA
Canine Triiodothyronine/T3 ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of canine Triiodothyronine/T3 in serum, plasma, tissue homogenates, and other biological fluids.
  • Ready-to-use ELISA kit
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%
Canine Triiodothyronine/T3 ELISA kit employs the competitive enzyme immunoassay technique for the quantitative measurement of canine Triiodothyronine/T3 in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with Triiodothyronine/T3 antigen. During the incubation, Triiodothyronine/T3 present in the samples or standards competes with the fixed amount of immobilised Triiodothyronine/T3 for binding sites on the Biotinylated Anti-Triiodothyronine/T3 Antibody. The more Triiodothyronine/T3 present in a sample or standard, the less Biotinylated Anti-Triiodothyronine/T3 Antibody that binds to the plate. Following incubation, unbound Biotinylated Anti-Triiodothyronine/T3 Antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of Triiodothyronine/T3 present in each sample or standard. The concentration of Triiodothyronine/T3 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.
Informations de commande: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité