Human Neuromedin B ELISA Kit
Fournisseur: ANTIBODIES.COM
A302906-96
ANTIA302906-96EA 630 EUR InStock
ANTIA302906-96
Human Neuromedin B ELISA Kit
Tests Tests ELISA
Human Neuromedin B ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Neuromedin B in serum, plasma, tissue homogenates, and other biological fluids.
  • Ready-to-use ELISA kit
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%
Human Neuromedin B ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Neuromedin B in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Neuromedin B has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Neuromedin B present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Neuromedin B Antibody, which binds the captured Neuromedin B present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Neuromedin B captured in each well. The concentration of Neuromedin B can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.
Informations de commande: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité