A303389-96
ANTIA303389-96EA
610
EUR
InStock
ANTIA303389-96
Mouse 25-Hydroxy Vitamin D ELISA Kit
Tests
Tests ELISA
Mouse 25-hydroxy vitamin D ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of mouse 25-hydroxy vitamin D in serum, plasma, tissue homogenates, and other biological fluids.
- Ready to use ELISA kit
- Assay precision: Intra assay: CV <8%, Inter assay: CV <10%
Mouse 25-Hydroxy vitamin D ELISA kit (A303389) employs the competitive enzyme immunoassay technique for the quantitative measurement of mouse 25-Hydroxy vitamin D in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with 25-Hydroxy vitamin D antigen. During the incubation, 25-Hydroxy vitamin D present in the samples or standards competes with the fixed amount of immobilised 25-Hydroxy vitamin D for binding sites on the Biotinylated anti-25-Hydroxy vitamin D antibody. The more 25-Hydroxy vitamin D present in a sample or standard, the less Biotinylated anti-25-Hydroxy vitamin D antibody that binds to the plate. Following incubation, unbound Biotinylated anti-25-Hydroxy vitamin D antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of 25-Hydroxy vitamin D present in each sample or standard. The concentration of 25-Hydroxy vitamin D can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.
Informations de commande: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.