Poly (ADP-Ribose) Polymer ELISA Kit
Fournisseur: ANTIBODIES.COM
A303880-96
ANTIA303880-96EA 610 EUR InStock
ANTIA303880-96
Poly (ADP-Ribose) Polymer ELISA Kit
Tests Tests ELISA
Poly (ADP-Ribose) Polymer ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of Poly (ADP-Ribose) Polymer in serum, plasma, tissue homogenates, and other biological fluids.
  • Ready to use ELISA kit
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%
Poly (ADP-Ribose) Polymer ELISA kit (A303880) employs the competitive enzyme immunoassay technique for the quantitative measurement of universal Poly (ADP-Ribose) Polymer in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with Poly (ADP-Ribose) Polymer antigen. During the incubation, Poly (ADP-Ribose) Polymer present in the samples or standards competes with the fixed amount of immobilised Poly (ADP-Ribose) Polymer for binding sites on the Biotinylated Anti-Poly (ADP-Ribose) Polymer Antibody. The more Poly (ADP-Ribose) Polymer present in a sample or standard, the less Biotinylated Anti-Poly (ADP-Ribose) Polymer Antibody that binds to the plate. Following incubation, unbound Biotinylated Anti-Poly (ADP-Ribose) Polymer Antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of Poly (ADP-Ribose) Polymer present in each sample or standard. The concentration of Poly (ADP-Ribose) Polymer can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.
Informations de commande: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité