15-HETE ELISA Kit
Fournisseur: ANTIBODIES.COM
A303889-96
ANTIA303889-96EA 610 EUR InStock
ANTIA303889-96
15-HETE ELISA Kit
Tests Tests ELISA
15-HETE ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of 15-HETE in serum, plasma, tissue homogenates, and other biological fluids.
  • Ready to use ELISA kit
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%
15-HETE ELISA kit (A303889) employs the competitive enzyme immunoassay technique for the quantitative measurement of universal 15-HETE in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with 15-HETE antigen. During the incubation, 15-HETE present in the samples or standards competes with the fixed amount of immobilised 15-HETE for binding sites on the Biotinylated anti-15-HETE antibody. The more 15-HETE present in a sample or standard, the less Biotinylated anti-15-HETE antibody that binds to the plate. Following incubation, unbound Biotinylated anti-15-HETE antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of 15-HETE present in each sample or standard. The concentration of 15-HETE can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.
Informations de commande: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité