A310382-96
ANTIA310382-96EA
525
EUR
InStock
ANTIA310382-96
Mouse Desmoglein 3/PVA ELISA Kit
Tests
Tests ELISA
Mouse Desmoglein 3/PVA ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Desmoglein 3/PVA in serum, plasma, and other biological fluids.
- Higher throughput: Get results in just 90 minutess, with a single wash step
- Detection Range: 0,75 to 12 ng/ml
- Sensitivity: 0,024 ng/ml
- Sample Type: Serum, plasma or other biological fluids
- Assay Precision: Intra-assay: CV <8%, Inter-assay: CV <10%
Mouse Desmoglein 3/PVA ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse Desmoglein 3/PVA in serum, plasma or other biological fluids. An antibody specific for Desmoglein 3/PVA has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Desmoglein 3/PVA present in each sample is bound to the wells by the immobilised antibody. Biotinylated anti-Desmoglein 3/PVA antibody, which also binds the Desmoglein 3/PVA present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-Desmoglein 3/PVA antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-Desmoglein 3/PVA antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of Desmoglein 3/PVA captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of Desmoglein 3/PVA in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.
Informations de commande: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, streptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.