A310965-96
ANTIA310965-96EA
545
EUR
InStock
ANTIA310965-96
Human HSD11B1 ELISA Kit
Tests
Tests ELISA
Human HSD11B1 ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human HSD11B1 in serum, plasma, and other biological fluids.
- Higher throughput: Get results in just 90 minutess, with a single wash step
- Detection Range: 300 to 4800 ng/L
- Sensitivity: 14,25 ng/L
- Sample Type: Serum, plasma or other biological fluids
- Assay Precision: Intra-assay: CV <8%, Inter-assay: CV <10%
Human HSD11B1 ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of human HSD11B1 in serum, plasma or other biological fluids. An antibody specific for HSD11B1 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the HSD11B1 present in each sample is bound to the wells by the immobilised antibody. Biotinylated anti-HSD11B1 antibody, which also binds the HSD11B1 present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-HSD11B1 antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-HSD11B1 antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of HSD11B1 captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of HSD11B1 in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.
Informations de commande: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, streptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.