Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Mouse SFRP5 ELISA Kit

Fournisseur: ANTIBODIES.COM

A311217-96
ANTIA311217-96EA 545 EUR InStock
ANTIA311217-96
Mouse SFRP5 ELISA Kit
Tests Tests ELISA
Mouse SFRP5 ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse SFRP5 in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Detection range: 300 to 4800 ng/ml
  • Sensitivity: 14,11 ng/ml
  • Sample type: Serum, plasma or other biological fluids
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%

Mouse SFRP5 ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse SFRP5 in serum, plasma or other biological fluids. An antibody specific for SFRP5 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the SFRP5 present in each sample is bound to the wells by the immobilised antibody. Biotinylated Anti-SFRP5 antibody, which also binds the SFRP5 present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-SFRP5 antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-SFRP5 antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. colour develops in proportion to the amount of SFRP5 captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of SFRP5 in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Informations de commande: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, streptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.


Commander maintenant

 

COMMANDE

SPÉCIFICATIONS

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité