A311827-96
ANTIA311827-96EA
545
EUR
InStock
ANTIA311827-96
Mouse hHR23b ELISA Kit
Tests
Tests ELISA
Mouse hHR23b ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse hHR23b in serum, plasma, and other biological fluids.
- Higher throughput: Get results in just 90 minutess, with a single wash step
- Detection range: 100 to 1600 ng/L
- Sensitivity: 7,05 ng/L
- Sample Type: Serum, plasma or other biological fluids
- Assay precision: Intra assay: CV <8%, Inter assay: CV <10%
Mouse hHR23b ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse hHR23b in serum, plasma or other biological fluids. An antibody specific for hHR23b has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the hHR23b present in each sample is bound to the wells by the immobilised antibody. Biotinylated anti-hHR23b antibody, which also binds the hHR23b present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-hHR23b antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-hHR23b antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. colour develops in proportion to the amount of hHR23b captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of hHR23b in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.
Informations de commande: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, steptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.