A311965-96
ANTIA311965-96EA
545
EUR
InStock
ANTIA311965-96
Human HDAC11/HD11 ELISA Kit
Tests
Tests ELISA
Human HDAC11/HD11 ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human HDAC11/HD11 in serum, plasma, and other biological fluids.
- Higher throughput: Get results in just 90 minutess, with a single wash step
- Detection range: 250 to 4000 ng/L
- Sensitivity: 1,17 ng/L
- Sample Type: Serum, plasma or other biological fluids
- Assay precision: Intra assay: CV <8%, Inter assay: CV <10%
Human HDAC11/HD11 ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of human HDAC11/HD11 in serum, plasma or other biological fluids. An antibody specific for HDAC11/HD11 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the HDAC11/HD11 present in each sample is bound to the wells by the immobilised antibody. Biotinylated anti-HDAC11/HD11 antibody, which also binds the HDAC11/HD11 present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-HDAC11/HD11 antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-HDAC11/HD11 antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. colour develops in proportion to the amount of HDAC11/HD11 captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of HDAC11/HD11 in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.
Informations de commande: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, steptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.