Human Calcium Pump PMCA2 ATPase ELISA Kit
Fournisseur: ANTIBODIES.COM
A313318-96
ANTIA313318-96EA 545 EUR InStock
ANTIA313318-96
Human Calcium Pump PMCA2 ATPase ELISA Kit
Tests Tests ELISA
Human Calcium Pump PMCA2 ATPase ELISA kit is a 90 minute sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Calcium Pump PMCA2 ATPase in serum, plasma, and other biological fluids.
  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%
Human Calcium Pump PMCA2 ATPase ELISA kit (A313318) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Calcium Pump PMCA2 ATPase in serum, plasma or other biological fluids. An antibody specific for Calcium Pump PMCA2 ATPase has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Calcium Pump PMCA2 ATPase present in each sample is bound to the wells by the immobilised antibody. Biotinylated anti-Calcium Pump PMCA2 ATPase antibody, which also binds the Calcium Pump PMCA2 ATPase present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-Calcium Pump PMCA2 ATPase antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-Calcium Pump PMCA2 ATPase antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of Calcium Pump PMCA2 ATPase captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of Calcium Pump PMCA2 ATPase in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.
Informations de commande: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, positive control, negative control, HRP-conjugated detection antibody, sample diluent, substrate solution A, substrate solution B, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité
Ce site utilise des cookies, en provenance de VWR ou de ses partenaires, afin de collecter des informations statistiques sur votre navigation et vous proposer des contenus en accord avec vos préférences, générés en fonction de vos habitudes de navigation. En poursuivant la consultation de ce site, vous approuvez l’utilisation de ces cookies.
Pour en savoir plus, consultez notre Politique de Confidentialit&eacute;..