Human KCP ELISA Kit
Fournisseur: ANTIBODIES.COM
A313517-96
ANTIA313517-96EA 545 EUR InStock
ANTIA313517-96
Human KCP ELISA Kit
Testen ELISAs
Human KCP ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human KCP in serum, plasma, and other biological fluids.
  • Higher throughput: Get results in just 90 minutess, with a single wash step
  • Detection range: 0,75 to 12 ng/ml
  • Sensitivity: 0,022 ng/ml
  • Sample type: Serum, plasma or other biological fluids
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%
Human KCP ELISA kit (A313517) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human KCP in serum, plasma or other biological fluids. An antibody specific for KCP has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the KCP present in each sample is bound to the wells by the immobilised antibody. Biotinylated anti-KCP antibody, which also binds the KCP present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-KCP antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-KCP antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of KCP captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of KCP in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.
Informations de commande: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, steptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité
Ce site utilise des cookies, en provenance de VWR ou de ses partenaires, afin de collecter des informations statistiques sur votre navigation et vous proposer des contenus en accord avec vos préférences, générés en fonction de vos habitudes de navigation. En poursuivant la consultation de ce site, vous approuvez l’utilisation de ces cookies.
Pour en savoir plus, consultez notre Politique de Confidentialit&eacute;..