A313560-96
ANTIA313560-96EA
545
EUR
InStock
ANTIA313560-96
Human Semenogelin I ELISA Kit
Tests
Tests ELISA
Human Semenogelin I ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Semenogelin I in serum, plasma, and other biological fluids.
- Higher throughput: Get results in just 90 minutes, with a single wash step
- Detection range: 3 to 48 ng/ml
- Sensitivity: 0,16 ng/ml
- Sample type: Serum, plasma or other biological fluids
- Assay precision: Intra assay: CV <8%, Inter assay: CV <10%
Human Semenogelin I ELISA kit (A313560) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Semenogelin I in serum, plasma or other biological fluids. An antibody specific for Semenogelin I has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Semenogelin I present in each sample is bound to the wells by the immobilised antibody. Biotinylated Anti-Semenogelin I antibody, which also binds the Semenogelin I present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-Semenogelin I antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-Semenogelin I antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of Semenogelin I captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of Semenogelin I in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.
Informations de commande: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, streptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.