A314034-96
ANTIA314034-96EA
545
EUR
InStock
ANTIA314034-96
Human CLMN ELISA Kit
Tests
Tests ELISA
Human CLMN ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human CLMN in serum, plasma, and other biological fluids.
- Higher throughput: Get results in just 90 minutess, with a single wash step
- Detection Range: 2,5 to 40 ng/ml
- Sensitivity: 0,12 ng/ml
- Sample Type: Serum, plasma or other biological fluids
- Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%
Human CLMN ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of human CLMN in serum, plasma or other biological fluids. An antibody specific for CLMN has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the CLMN present in each sample is bound to the wells by the immobilised antibody. Biotinylated anti-CLMN antibody, which also binds the CLMN present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-CLMN antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-CLMN antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. colour develops in proportion to the amount of CLMN captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of CLMN in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.
Informations de commande: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, steptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.