Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

5-Hydroxyindoleacetic Acid ELISA Kit

Fournisseur: ANTIBODIES.COM

A74044-96
ANTIA74044-96EA 600 EUR InStock
ANTIA74044-96
5-Hydroxyindoleacetic Acid ELISA Kit
Tests Tests ELISA
5-Hydroxyindoleacetic Acid ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of 5-Hydroxyindoleacetic Acid in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-to-use ELISA kit
  • Detection Range: 1,563 to 100 ng/ml
  • Sensitivity: 0,938 ng/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%

5-Hydroxyindoleacetic Acid ELISA Kit employs the competitive enzyme immunoassay technique for the quantitative measurement of universal 5-Hydroxyindoleacetic Acid in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with 5-Hydroxyindoleacetic Acid antigen. During the incubation, 5-Hydroxyindoleacetic Acid present in the samples or standards competes with the fixed amount of immobilised 5-Hydroxyindoleacetic Acid for binding sites on the Biotinylated Anti-5-Hydroxyindoleacetic Acid Antibody. The more 5-Hydroxyindoleacetic Acid present in a sample or standard, the less Biotinylated Anti-5-Hydroxyindoleacetic Acid Antibody that binds to the plate. Following incubation, unbound Biotinylated Anti-5-Hydroxyindoleacetic Acid Antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of 5-Hydroxyindoleacetic Acid present in each sample or standard. The concentration of 5-Hydroxyindoleacetic Acid can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Informations de commande: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.


Commander maintenant

 

COMMANDE

SPÉCIFICATIONS

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité