A76997-96
ANTIA76997-96EA
580
EUR
InStock
ANTIA76997-96
Mouse Mup20 ELISA Kit
Tests
Tests ELISA
Mouse Mup20 ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of mouse Mup20 in serum, plasma, tissue homogenates, and other biological fluids.
- Ready to use ELISA kit
- Detection range: 0,313 to 20 ng/ml
- Sensitivity: 0,188 ng/ml
- Sample type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay precision: Intra assay: CV <8%, Inter assay: CV <10%
Mouse Mup20 employs the competitive enzyme immunoassay technique for the quantitative measurement of mouse Mup20 in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with Mup20 antigen. During the incubation, Mup20 present in the samples or standards competes with the fixed amount of immobilised Mup20 for binding sites on the Biotinylated Anti-Mup20 Antibody. The more Mup20 present in a sample or standard, the less Biotinylated Anti-Mup20 Antibody that binds to the plate. Following incubation, unbound Biotinylated Anti-Mup20 Antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of Mup20 present in each sample or standard. The concentration of Mup20 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.
Informations de commande: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin Conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.