A78745-96
ANTIA78745-96EA
580
EUR
InStock
ANTIA78745-96
Human RPIA/PRI ELISA Kit
Tests
Tests ELISA
Human RPIA/PRI ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of human RPIA/PRI in serum, plasma, tissue homogenates, and other biological fluids.
- Ready To Use ELISA kit
- Detection Range: 1,563 to 100 ng/ml
- Sensitivity: 0,938 ng/ml
- Sample Type: Serum, plasma, tissue homogenates and other biological fluids
- Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%
Human RPIA/PRI ELISA kit (A78745) employs the competitive enzyme immunoassay technique for the quantitative measurement of human RPIA/PRI in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with RPIA/PRI antigen. During the incubation, RPIA/PRI present in the samples or standards competes with the fixed amount of immobilized RPIA/PRI for binding sites on the Biotinylated Anti-RPIA/PRI Antibody. The more RPIA/PRI present in a sample or standard, the less Biotinylated Anti-RPIA/PRI Antibody that binds to the plate. Following incubation, unbound Biotinylated Anti-RPIA/PRI Antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of RPIA/PRI present in each sample or standard. The concentration of RPIA/PRI can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.
Informations de commande: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.