A78937-96
ANTIA78937-96EA
580
EUR
InStock
ANTIA78937-96
Mouse Prealbumin ELISA Kit
Tests
Tests ELISA
Mouse Prealbumin ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Prealbumin in serum, plasma, tissue homogenates, and other biological fluids.
- Ready-To-Use ELISA Kit
- Detection Range: 0,781 - 50 ng/ml
- Sensitivity: 1,875 ng/ml
- Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%
Mouse Prealbumin ELISA kit (A78937) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse Prealbumin in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Prealbumin has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Prealbumin present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Prealbumin Antibody, which binds the captured Prealbumin present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Prealbumin captured in each well. The concentration of Prealbumin can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.
Informations de commande: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin Conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.