A79018-96
ANTIA79018-96EA
700
EUR
InStock
ANTIA79018-96
Monkey Factor VIII ELISA Kit
Tests
Tests ELISA
Monkey Factor VIII ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of monkey Factor VIII in serum, plasma, tissue homogenates, and other biological fluids.
- Ready To Use ELISA kit
- Detection Range: 2,344 to 150 ng/ml
- Sensitivity: 1,406 ng/ml
- Sample Type: Serum, plasma, tissue homogenates and other biological fluids
- Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%
Monkey Factor VIII ELISA kit (A79018) employs the sandwich enzyme immunoassay technique for the quantitative measurement of monkey Factor VIII in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Factor VIII has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Factor VIII present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Factor VIII Antibody, which binds the captured Factor VIII present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Factor VIII captured in each well. The concentration of Factor VIII can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.
Informations de commande: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.