Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Rat Apolipoprotein A II/ApoA-II ELISA Kit

Fournisseur: ANTIBODIES.COM

A79117-96
ANTIA79117-96EA 545 EUR InStock
ANTIA79117-96
Rat Apolipoprotein A II/ApoA-II ELISA Kit
Tests Tests ELISA
Rat Apolipoprotein A II/ApoA-II ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat Apolipoprotein A II/ApoA-II in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready To Use ELISA Kit
  • Detection Range: 6,25 to 400 ng/ml
  • Sensitivity: 3,75 ng/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%

Rat Apolipoprotein A II / ApoA-II ELISA Kit (A79117) employs the sandwich enzyme immunoassay technique for the quantitative measurement of rat Apolipoprotein A II / ApoA-II in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Apolipoprotein A II / ApoA-II has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Apolipoprotein A II / ApoA-II present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Apolipoprotein A II / ApoA-II Antibody, which binds the captured Apolipoprotein A II / ApoA-II present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Apolipoprotein A II / ApoA-II captured in each well. The concentration of Apolipoprotein A II / ApoA-II can then be calculated by reading the O.D. absorbance at 450nm in a microplate reader and referring to the standard curve.

Informations de commande: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.


Commander maintenant

 

COMMANDE

SPÉCIFICATIONS

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité