A79816-96
ANTIA79816-96EA
600
EUR
InStock
ANTIA79816-96
Human Pregnanediol-3-Glucuronide ELISA Kit
Tests
Tests ELISA
Human Pregnanediol-3-Glucuronide ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of human Pregnanediol-3-Glucuronide in serum, plasma, tissue homogenates, and other biological fluids.
- Ready To Use ELISA kit
- Detection Range: 15,625 to 1000 pg/ml
- Sensitivity: 9,375 pg/ml
- Sample Type: Serum, plasma, tissue homogenates and other biological fluids
- Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%
Human Pregnanediol-3-Glucuronide ELISA kit (A79816) employs the competitive enzyme immunoassay technique for the quantitative measurement of human Pregnanediol-3-Glucuronide in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with Pregnanediol-3-Glucuronide antigen. During the incubation, Pregnanediol-3-Glucuronide present in the samples or standards competes with the fixed amount of immobilized Pregnanediol-3-Glucuronide for binding sites on the Biotinylated Anti-Pregnanediol-3-Glucuronide Antibody. The more Pregnanediol-3-Glucuronide present in a sample or standard, the less Biotinylated Anti-Pregnanediol-3-Glucuronide Antibody that binds to the plate. Following incubation, unbound Biotinylated Anti-Pregnanediol-3-Glucuronide Antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of Pregnanediol-3-Glucuronide present in each sample or standard. The concentration of Pregnanediol-3-Glucuronide can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.
Informations de commande: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.