Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Human p27 KIP 1 ELISA Kit

Fournisseur: ANTIBODIES.COM

A87118-96
ANTIA87118-96EA 600 EUR InStock
ANTIA87118-96
Human p27 KIP 1 ELISA Kit
Tests Tests ELISA
Human p27 KIP 1 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human p27 KIP 1 in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-to-use ELISA Kit
  • Detection range: 0,156 to 10 ng/ml
  • Sensitivity: 0,094 ng/ml
  • Sample type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%

Human p27 KIP 1 ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of human p27 KIP 1 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for p27 KIP 1 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the p27 KIP 1 present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-p27 KIP 1 Antibody, which binds the captured p27 KIP 1 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of p27 KIP 1 captured in each well. The concentration of p27 KIP 1 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Informations de commande: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin Conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.


Commander maintenant

 

COMMANDE

SPÉCIFICATIONS

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité