A87276-96
ANTIA87276-96EA
580
EUR
InStock
ANTIA87276-96
Rat 8-Hydroxydeoxyguanosine ELISA Kit
Tests
Tests ELISA
Rat 8-Hydroxydeoxyguanosine ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of rat 8-Hydroxydeoxyguanosine in serum, plasma, tissue homogenates, and other biological fluids.
- Ready to use ELISA kit
- Detection range: 1,563 to 100 ng/ml
- Sensitivity: 0,938 ng/ml
- Sample type: Serum, plasma, tissue homogenates and other biological fluids
- Assay precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%
Rat 8-Hydroxydeoxyguanosine ELISA kit employs the competitive enzyme immunoassay technique for the quantitative measurement of rat 8-Hydroxydeoxyguanosine in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with 8-Hydroxydeoxyguanosine antigen. During the incubation, 8-Hydroxydeoxyguanosine present in the samples or standards competes with the fixed amount of immobilised 8-Hydroxydeoxyguanosine for binding sites on the Biotinylated Anti-8-Hydroxydeoxyguanosine Antibody. The more 8-Hydroxydeoxyguanosine present in a sample or standard, the less Biotinylated Anti-8-Hydroxydeoxyguanosine Antibody that binds to the plate. Following incubation, unbound Biotinylated Anti-8-Hydroxydeoxyguanosine Antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of 8-Hydroxydeoxyguanosine present in each sample or standard. The concentration of 8-Hydroxydeoxyguanosine can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.
Informations de commande: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.