Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Bovine Natriuretic Peptides A ELISA Kit

Fournisseur: ANTIBODIES.COM

A87432-96
ANTIA87432-96EA 700 EUR InStock
ANTIA87432-96
Bovine Natriuretic Peptides A ELISA Kit
Tests Tests ELISA
Bovine Natriuretic Peptides A ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of bovine Natriuretic Peptides A in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-to-use ELISA Kit
  • Detection range: 15,625 to 1000 pg/ml
  • Sensitivity: 9,375 pg/ml
  • Sample type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%

Bovine Natriuretic Peptides A ELISA kit employs the competitive enzyme immunoassay technique for the quantitative measurement of bovine Natriuretic Peptides A in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with Natriuretic Peptides A antigen. During the incubation, Natriuretic Peptides A present in the samples or standards competes with the fixed amount of immobilised Natriuretic Peptides A for binding sites on the Biotinylated Anti-Natriuretic Peptides A Antibody. The more Natriuretic Peptides A present in a sample or standard, the less Biotinylated Anti-Natriuretic Peptides A Antibody that binds to the plate. Following incubation, unbound Biotinylated Anti-Natriuretic Peptides A Antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of Natriuretic Peptides A present in each sample or standard. The concentration of Natriuretic Peptides A can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Informations de commande: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin Conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.


Commander maintenant

 

COMMANDE

SPÉCIFICATIONS

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité