Synonyms:
JNK Inhibitor II, Anthra[1,9-cd]pyrazol-6(2H)-one, Dibenzo[cd,g]indazol-6(2H)-one, SP600125, SP 600125
BIS1100-1G
BIS1100-300MG
APOSBIS1100-1GEA
444
EUR
InStock
APOSBIS1100-1G
APOSBIS1100-300MG
1,9-Pyrazoloanthrone ≥99%
1,9-Pyrazoloanthrone
Novel, potent and selective JNK-1,-2, and -3 inhibitor (Ki = 0.19 uM). SP600125 is an ATP-competitive reversible inhibitor with >20-fold selectivity vs. a range of kinases and enzymes tested. Proc. Natl. Acad. Sci. USA 98: 13681-13686 (2001).