Synonyms:
3-[(Methylsulphonyl)sulphanyl]propionic acid, 2-Carboxyethyl methylthiosulphonate
OR7810T-100MG
APOSOR7810T-100MGEA
284
EUR
InStock
APOSOR7810T-100MG
3-[(Methylsulphonyl)thio]propanoic acid
3-[(Methylsulphonyl)thio]propanoic acid
Reacts specifically and rapidly with thiols to form mixed disulfides. Useful for probing the structures & functions of receptor & ion channel proteins.