PC5740-1KG
APOSPC5740-1KGEA
0
EUR
InStock
APOSPC5740-1KG
Acide pentafluoropropionique 98%
Acide pentafluoropropionique
The K salt is an alternative to iodopentafluoroethane for conversion of aryl bromides and iodides to corresponding pentafluoroethyl derivatives without sealed tube chemistry.