DrySyn® MAXI Heating Blocks, 2 L
Fournisseur: ASYNT
ADS2-2
ASYNADS2-2EA 0 EUR InStock
ASYNADS2-2
DrySyn® MAXI Heating Blocks, 2 L
Chauffe-blocs Blocs pour bains
The DrySyn® MAXI 2 L is a stand-alone 2000 ml version of the standard DrySyn MAXI Kit (which alternatively has a 3000 ml base and 2000 ml insert) which comes with heat resistant safety lifting handles.
  • Enables clean, safe synthesis without the hazards or problems associated with silicon oil baths or heating mantles
  • With no messy oil, heat resistant handles, and simple to configure range of inserts this system is world renowned
  • With heating rates the same as an oil bath there is no need to change your chemistry either
  • Fits standard hotplate stirrers with ease
Does not require any further base, and is suitable for heating 2000 ml round bottom flasks.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité