B20021.09
B20021.09EA
41.8
EUR
InStock
B20021.09
Azoxybenzène ≥98%
Azoxybenzene
Purity: 98+%
CAS No.: 495-48-7
Molecular Formula: C12H10N2O
Formula Weight: 198.23
Melting Point: 33-36°
Boiling Point: 130°/0.9mm
Merck Reference: 14,923
MDL No.: MFCD00019925