Synonyms:
Perfluorohexyl iodide, Perfluoro-n-hexyl iodide, 1,1,1,2,2,3,3,4,4,5,5,6,6-Tridecafluoro-6-iodohexane, Perfluoro-1-iodohexane
Stabilization:
Stabilised with copper
B20958.18
B20958.18EA
164
EUR
InStock
B20958.18
1-Iodotridécafluorohexane 98+% stabilisé
1-Iodotridécafluorohexane
Purity: 98+%
CAS No.: 355-43-1
Molecular Formula: CF3(CF2)5I
Formula Weight: 445.95
Refractive Index: 1.3290
Melting Point: -45°
Boiling Point: 117°
Beilstein Reference: 1802118
MDL No.: MFCD00001063