B21209.14
B21209.14EA
99.1
EUR
InStock
B21209.14
1-Chloro-2-fluorobenzène ≥98%
1-Chloro-2-fluorobenzène
Purity: 98+%
CAS No.: 348-51-6
Molecular Formula: C6H4ClF
Formula Weight: 130.55
Flash Point: 31°(87°F)
Refractive Index: 1.5010
Melting Point: -43°
Boiling Point: 137-138°
Beilstein Reference: 1855301
UN No.: 1993
MDL No.: MFCD00000533