Synonyms:
5-Hydroxytryptamine Hydrochloride
B21263.03
B21263.06
B21263.09
B21263.A1
B21263.03EA
122
EUR
InStock
B21263.03
B21263.06
B21263.09
B21263.A1
3-(2-Aminoéthyl)-5-hydroxyindole chlorhydrate ≥98%
3-(2-Aminoéthyl)-5-hydroxyindole chlorhydrate
Purity: 98%
CAS No.: 153-98-0
Molecular Formula: C10H12N2O.HCl
Formula Weight: 212.68
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Sensitivity: Light Sensitive and Hygroscopic
Appearance: White to pale cream
Melting Point: 167-172°
Beilstein Reference: 3570963
Merck Reference: 14,8463
UN No.: 2811
MDL No.: MFCD00012686